Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Trimethylsilyl)imidazole

Catalog No. 92751250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-751-250ML 250 mL
92-751-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 92-751-250ML Supplier Sigma Aldrich Supplier No. 92751250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-(trimethylsilyl)imidazole may be used as an effective alternative to n,o-bis(trimethylsilyl)trifluoroacetamide (bstfa) and n-methyl-n-(trimethylsilyl)trifluoroacetamide (mstfa) for the efficient silylation of ti-mcm-41, a titanium substituted mesoporous molecular sieve
  • This process enhances the hydrophobicity and catalytic activity of ti-mcm-41 catalyst for cyclohexene epoxidation with aqueous hydrogen peroxide.powerful silylating agent, particularly for alcohols; synthesis of acyl imidazolides

Specifications

CAS 18156-74-6
Density 0.957 g/mL at 20°C, 0.956 g/mL (at 25°C (literature))
Boiling Point 93°C to 94°C (14 mmHg)
Molecular Formula C6H12N2Si
Refractive Index n20/D 1.475 (literature)
Linear Formula C6H12N2Si
MDL Number MFCD00005280
Quantity 250 mL
Synonym N-Trimethylsilylimidazole; TMSI; TSIM
Molecular Weight (g/mol) 140.26
Percent Purity ≥98.0% (T); ≥98.0%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.