Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trimethylsilyl)imidazole - Pyridine Mixture, For GC derivatization, MilliporeSigma™ Supelco™

Catalog No. 111018769 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8769 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8769 Supplier MilliporeSigma Supelco Supplier No. 9271810X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00284770
Synonym Silylating mixture VII; TMSI+Pyridine

Specifications

CAS 8077-35-8
Density 0.98 g/mL (at 20°C)
Flash Point 17°C
Quantity 10 x 1 mL
Grade For GC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.