Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Trimethylsilyl)propyne

Catalog No. 2444815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-448-15G 5 g
24-448-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-448-15G Supplier Sigma Aldrich Supplier No. 2444815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(Trimethylsilyl)propyne was used in the synthesis of highly substituted indenes via palladium-catalyzed carboannulation and indenones via a rhodium-catalyzed reaction with 2-bromophenylboronic acids.

Specifications

CAS 6224-91-5
Density 0.758 g/mL (at 25°C (literature))
Boiling Point 99°C to 100°C (lit.)
Molecular Formula C6H12Si
Refractive Index n20/D 1.417 (literature)
Linear Formula CH3C///CSi(CH3)3
MDL Number MFCD00009271
Quantity 5 g
Synonym Trimethyl-1-propynylsilane
Molecular Weight (g/mol) 112.24
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.