Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Triphenylphosphoranylidene-2-propanone, 99%

Catalog No. AC169970250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169970250 25 g Glass bottle
AC169971000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169970250 Supplier Thermo Scientific Chemicals Supplier No. 169970250
Add to Cart

CAS: 1439-36-7 | C21H19OP | 318.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1439-36-7
Molecular Formula C21H19OP
Molecular Weight (g/mol) 318.36
MDL Number MFCD00008774
InChI Key KAANTNXREIRLCT-UHFFFAOYSA-N
Synonym acetylmethylene triphenylphosphorane, 1-triphenylphosphoranylidene propan-2-one, 1-triphenylphosphoranylidene-2-propanone, 1-triphenylphosphoranylidene acetone, 2-propanone, 1-triphenylphosphoranylidene, acetonylidenetriphenyl phosphorane, acetylmethylene triphenylphosphine, triphenylacetylmethylene phosphorane, 2-propanone, triphenylphosphoranylidene, triphenylphosphoranylidene-2-propanone
PubChem CID 15038
IUPAC Name 1-(triphenyl-$l^{5}-phosphanylidene)propan-2-one
SMILES CC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 203.0°C to 207.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCOCH3
Quantity 25 g
Formula Weight 318.34
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1-Triphenylphosphoranylidene-2-propanone, 99%
Show More Show Less
EINECSNumber 215-878-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.