Learn More
Medchemexpress LLC 1-Trityl-1H-imidazole-4-carbaldehyde | 33016-47-6 | MFCD02179554 | 99.1% | C23H18N2O | 25 G
SDP
Catalog No. 5000358544
Encompass_Preferred
1-Trityl-1H-imidazole-4-carbaldehyde is a biochemical reagent used in life science research. It can be utilized as a biological material or organic compound for various research applications. This product is for research use only and not intended for patient use.
  • Appears as a white to off-white solid
  • Used in the synthesis of histamine and girolline
Catalog No. 50-003-58544 Supplier Medchemexpress LLC Supplier No. 25GHY77049
1-Trityl-1H-imidazole-4-carbaldehyde is a biochemical reagent used in life science research. It can be utilized as a biological material or organic compound for various research applications. This product is for research use only and not intended for patient use.
  • Appears as a white to off-white solid
  • Used in the synthesis of histamine and girolline