Learn More
VAZYME INTERNATIONAL LLC Vazyme SupRealQ Ultra Hunter SYBR qPCR Master Mix (U+), UDG System, High Precision
Catalog No. NC3979689
Encompass
This specialized premix for qPCR reactions uses the SYBR Green I fluorescence method and features a purple color for easy sample loading. It includes a Taq polymerase enzyme for high specificity and a hot-start function. A dUTP/UDG contamination prevention system operates at room temperature, and a special ROX Passive Reference Dye ensures compatibility with various qPCR instruments.
  • Designed for high precision with low-abundance targets.
  • Handles degraded and high-GC templates effectively.
  • Features dUTP/heat-labile UDG system for room temperature contaminant elimination.
  • Pre-mixed formula offers consistent performance and 72-hour stability.
  • Ensures stable amplification of complex templates.
  • Exhibits 90–110% amplification efficiency across a wide linear range.
  • Efficient anti-contamination, with over 99.99% removal.
Catalog No. NC3979689 Supplier VAZYME INTERNATIONAL LLC Supplier No. Q71302
This specialized premix for qPCR reactions uses the SYBR Green I fluorescence method and features a purple color for easy sample loading. It includes a Taq polymerase enzyme for high specificity and a hot-start function. A dUTP/UDG contamination prevention system operates at room temperature, and a special ROX Passive Reference Dye ensures compatibility with various qPCR instruments.
  • Designed for high precision with low-abundance targets.
  • Handles degraded and high-GC templates effectively.
  • Features dUTP/heat-labile UDG system for room temperature contaminant elimination.
  • Pre-mixed formula offers consistent performance and 72-hour stability.
  • Ensures stable amplification of complex templates.
  • Exhibits 90–110% amplification efficiency across a wide linear range.
  • Efficient anti-contamination, with over 99.99% removal.