Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Undecanol | 112-42-5 | 172.31 | 1 G
SDP

Catalog No. 5000449205
Encompass_Preferred

1-Undecanol, also known as Undecyl alcohol, is primarily formed from the degradation of 2-tridecanone by Pseudomonas bacteria. This compound has been observed to enhance the attraction of *Grapholita molesta* to sex pheromone traps, acting as a synergist.

  • Main product from bacterial degradation of 2-tridecanone
  • Enhances attraction to sex pheromone traps
  • Acts as a synergist for *Grapholita molesta* sex pheromone
  • Appearance is solid below 11°C and liquid above 11°C
  • Colorless to light yellow
  • Soluble in DMSO

Catalog No. 50-004-49205 Supplier Medchemexpress LLC Supplier No. HYW0042921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Undecanol, also known as Undecyl alcohol, is primarily formed from the degradation of 2-tridecanone by Pseudomonas bacteria. This compound has been observed to enhance the attraction of *Grapholita molesta* to sex pheromone traps, acting as a synergist.

  • Main product from bacterial degradation of 2-tridecanone
  • Enhances attraction to sex pheromone traps
  • Acts as a synergist for *Grapholita molesta* sex pheromone
  • Appearance is solid below 11°C and liquid above 11°C
  • Colorless to light yellow
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.