Learn More
Medchemexpress LLC 1-Undecanol | 112-42-5 | 172.31 | 10 MM * 1 ML
SDP
Catalog No. 5000446935
Encompass_Preferred
1-Undecanol is primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from soil. It has been shown to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from soil.
  • Enhances the attraction of Grapholita molesta to sex pheromone traps.
  • Acts as a synergist for the sex pheromone of Grapholita molesta.
Catalog No. 50-004-46935 Supplier Medchemexpress LLC Supplier No. HYW00429210MM/1ML
1-Undecanol is primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from soil. It has been shown to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from soil.
  • Enhances the attraction of Grapholita molesta to sex pheromone traps.
  • Acts as a synergist for the sex pheromone of Grapholita molesta.