Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Undecanol | 112-42-5 | 172.31 | 5 G
SDP

Catalog No. 5000445419
Encompass_Preferred

1-Undecanol, also known as Undecyl alcohol, is a metabolite primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has shown potential in enhancing the attraction of Grapholita molesta to sex pheromone traps. It appears as a solid below 11°C and a liquid above 11°C, with a colorless to light yellow appearance.

  • Formula: C11H24O
  • Initial source: Plants other families and microorganisms (a strain of Pseudomonas)
  • Solubility: 100 mg/mL in DMSO (580.35 mM)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • For research use only

Catalog No. 50-004-45419 Supplier Medchemexpress LLC Supplier No. HYW0042925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Undecanol, also known as Undecyl alcohol, is a metabolite primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has shown potential in enhancing the attraction of Grapholita molesta to sex pheromone traps. It appears as a solid below 11°C and a liquid above 11°C, with a colorless to light yellow appearance.

  • Formula: C11H24O
  • Initial source: Plants other families and microorganisms (a strain of Pseudomonas)
  • Solubility: 100 mg/mL in DMSO (580.35 mM)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.