Learn More
Medchemexpress LLC 1-Undecanol-d23 | 349553-88-4 | 195.45 | 100 MG
SDP
Catalog No. 5000448848
Encompass_Preferred
1-Undecanol-d23 (Undecyl alcohol-d23) is the deuterium labeled 1-Undecanol. It is the main product generated from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from the soil. This compound can enhance the attraction of Grapholita molesta to sex pheromone traps. Deuterium (2H, D) has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-48848 Supplier Medchemexpress LLC Supplier No. HYW004292S100MG
Add to Cart
Add to Cart
1-Undecanol-d23 (Undecyl alcohol-d23) is the deuterium labeled 1-Undecanol. It is the main product generated from the degradation of 2-tridecanone by Pseudomonas bacteria isolated from the soil. This compound can enhance the attraction of Grapholita molesta to sex pheromone traps. Deuterium (2H, D) has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS