Learn More
Medchemexpress LLC 1-Undecanol-d23 | 349553-88-4 | 195.45 | 25 MG
SDP
Catalog No. 5000447040
Encompass_Preferred
1-Undecanol-d23 (Undecyl alcohol-d23) is a deuterium labeled version of 1-Undecanol. This compound is known to be generated from the degradation of 2-tridecanone by *Pseudomonas* bacteria. Deuterium labeled compounds are valuable tools in research, particularly in drug development, where they can serve as tracers and internal standards for various analytical methods. The incorporation of stable heavy isotopes can influence the pharmacokinetic and metabolic profiles of drug molecules.
  • Deuterium labeled compound
  • Functions as a tracer in research applications
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Generated from the degradation of 2-tridecanone
  • May enhance the attraction of *Grapholita molesta* to sex pheromone traps
  • Can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-47040 Supplier Medchemexpress LLC Supplier No. 25MGHYW004292S
Add to Cart
Add to Cart
1-Undecanol-d23 (Undecyl alcohol-d23) is a deuterium labeled version of 1-Undecanol. This compound is known to be generated from the degradation of 2-tridecanone by *Pseudomonas* bacteria. Deuterium labeled compounds are valuable tools in research, particularly in drug development, where they can serve as tracers and internal standards for various analytical methods. The incorporation of stable heavy isotopes can influence the pharmacokinetic and metabolic profiles of drug molecules.
  • Deuterium labeled compound
  • Functions as a tracer in research applications
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Generated from the degradation of 2-tridecanone
  • May enhance the attraction of *Grapholita molesta* to sex pheromone traps
  • Can affect pharmacokinetic and metabolic profiles of drugs