Learn More
Medchemexpress LLC 1-Undecanol-d23 | 349553-88-4 | 195.45 | 5 MG
SDP
Catalog No. 5000448912
Encompass_Preferred
1-Undecanol-d23 (Undecyl alcohol-d23) is a deuterium-labeled variant of 1-Undecanol, which is primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. It has been noted to enhance the attraction of Grapholita molesta to sex pheromone traps. Stable heavy isotopes, including deuterium, are incorporated into drug molecules for use as tracers in drug development and have the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-48912 Supplier Medchemexpress LLC Supplier No. HYW004292S5MG
Add to Cart
Add to Cart
1-Undecanol-d23 (Undecyl alcohol-d23) is a deuterium-labeled variant of 1-Undecanol, which is primarily produced from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. It has been noted to enhance the attraction of Grapholita molesta to sex pheromone traps. Stable heavy isotopes, including deuterium, are incorporated into drug molecules for use as tracers in drug development and have the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS