Learn More
Medchemexpress LLC 1-Undecanol (Undecyl alcohol) | 112-42-5 | 172.31 | 100 G
SDP
Catalog No. 5000445892
Encompass_Preferred
1-Undecanol (Undecyl alcohol) is primarily produced from bacteria through the oxidation of 2-tridecanone. It is also the main product resulting from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has been observed to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Main product generated from the degradation of 2-tridecanone by Pseudomonas bacteria.
  • Enhances attraction of Grapholita molesta to sex pheromone traps.
  • Acts as a synergist for Grapholita molesta sex pheromone at 50-200 μg doses, with 50 μg showing the most significant effect.
Catalog No. 50-004-45892 Supplier Medchemexpress LLC Supplier No. HYW004292100G
1-Undecanol (Undecyl alcohol) is primarily produced from bacteria through the oxidation of 2-tridecanone. It is also the main product resulting from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has been observed to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Main product generated from the degradation of 2-tridecanone by Pseudomonas bacteria.
  • Enhances attraction of Grapholita molesta to sex pheromone traps.
  • Acts as a synergist for Grapholita molesta sex pheromone at 50-200 μg doses, with 50 μg showing the most significant effect.