Learn More
Medchemexpress LLC 1-Undecanol (Undecyl alcohol) | 112-42-5 | 172.31 | 500 G
SDP
Catalog No. 5000447674
Encompass_Preferred
1-Undecanol, also known as Undecyl alcohol, is a metabolite primarily generated from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has demonstrated the ability to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Main product of 2-tridecanone degradation by Pseudomonas bacteria
  • Enhances attraction of Grapholita molesta to sex pheromone traps
  • Acts as a synergist for Grapholita molesta sex pheromone
Catalog No. 50-004-47674 Supplier Medchemexpress LLC Supplier No. HYW004292500G
1-Undecanol, also known as Undecyl alcohol, is a metabolite primarily generated from the degradation of 2-tridecanone by Pseudomonas bacteria found in soil. This compound has demonstrated the ability to enhance the attraction of Grapholita molesta to sex pheromone traps.
  • Main product of 2-tridecanone degradation by Pseudomonas bacteria
  • Enhances attraction of Grapholita molesta to sex pheromone traps
  • Acts as a synergist for Grapholita molesta sex pheromone