Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Undecene, 94.0%, For GC analysis, MP Biomedicals™

Catalog No. ICN15823805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15823805 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15823805 Supplier MP Biomedicals, Inc Supplier No. 0215823805
Add to Cart
Add to Cart

Chemical Identifiers

CAS 821-95-4
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
PubChem CID 13190
ChEBI CHEBI:77444
IUPAC Name undec-1-ene
SMILES CCCCCCCCCC=C

Specifications

Color Colorless
Assay Percent Range 0.94
Quantity 5 mL
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material 1-Undecene, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.