Learn More
Drew Scientific Inc MULTI-TROL™ Mouse Control, Calibration & Quality Control, 1 Vial
SDP
Catalog No. NC2338826
Encompass
The MULTI-TROL™ Mouse Control is a designed for calibration and quality control of veterinary hematology systems. Presented in a single 2 mL vial, it helps ensure the accuracy and reliability of hematological measurements for mouse samples. Regular usage, advised daily, enhances instrument performance, distinguishing it from other controls with its specific focus on mouse specimens.
Catalog No. NC2338826 Supplier Drew Scientific Inc Supplier No. 600065L
The MULTI-TROL™ Mouse Control is a designed for calibration and quality control of veterinary hematology systems. Presented in a single 2 mL vial, it helps ensure the accuracy and reliability of hematological measurements for mouse samples. Regular usage, advised daily, enhances instrument performance, distinguishing it from other controls with its specific focus on mouse specimens.