Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Vinyl-2-pyrrolidinone

Catalog No. 503126047 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6047 1 kg
50-312-5816 250 g
50-312-6048 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-6047 Supplier Sigma Aldrich Supplier No. V34091KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Vinyl-2-pyrrolidinone is a colorless to yellow liquid, with a characteristic odor. Its melting point is around 13.5oC and boils at about 90-92oC. VP is completely miscible in water and in most organic solvents but partially miscible in aliphatic hydrocarbons. Industrial production of VP by reacting 2-pyrrolidone with acetylene at high pressure and temperature has been reported. The vinylation process proceeds in liquid phase and is catalyzed by 3-pyrrolidone -potassium hydroxide.1-Vinyl-2-pyrrolidinone (VP) is used in the study of macrophotoinitiator characteristics of poly (vinylchloride) type molecules. Also, used to study the genotoxicity of polyvinylpyrrolidone coated silver nanoparticles.

Specifications

CAS 88-12-0
Density 1.04 g/mL (at 25°C)
Boiling Point 92°C to 95°C (11 mmHg)
Molecular Formula C6H9NO
Refractive Index n20/D 1.512 (literature)
Linear Formula C6H9NO
MDL Number MFCD00003197
Quantity 1 kg
Synonym 1-Vinyl-2-pyrrolidone
Molecular Weight (g/mol) 111.14
Percent Purity ≥99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.