Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Vinyl-2-pyrrolidinone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015740 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5740 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5740 Supplier MilliporeSigma Supelco Supplier No. PHR1740200MG
Add to Cart
Add to Cart

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 685°F
CAS 88-12-0
Vapor Pressure 0.1 mmHg (24°C)
Vapor Density 3.8 (vs. air)
Density 1.04 g/mL (at 25°C)
Linear Formula C6H9 NO
MDL Number MFCD00003197
Boiling Point 92°C to 95°C (11 mmHg)
Synonym 1-Vinyl-2-pyrrolidinone; 1-Vinyl-2-pyrrolidone
Type Certified Reference Material
Quantity 200 mg
Formula Weight 111.14
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.