Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Vinylimidazole

Catalog No. 235466100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-546-6100G 100 g
23-546-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-546-6100G Supplier Sigma Aldrich Supplier No. 235466100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-vinylimidazole has been employed:
  • As functional monomer in the synthesis of molecularly imprinted polymers
  • As monomer in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) [m-poly(dvb-vim)] microbeads
  • In the synthesis of novel ion imprinted polyvinylimidazole-silica hybrid copolymer (iihc)
  • In removal of the vinyl protecting group via isoprene-catalyzed lithiation process

Specifications

CAS 1072-63-5
Density 1.039 g/mL (at 25°C (literature))
Boiling Point 192°C to 194°C (lit.), 78°C to 79°C (13 mmHg)
Molecular Formula C5H6N2
Refractive Index n20/D 1.533 (literature)
Linear Formula C5H6N2
MDL Number MFCD00005297
Quantity 100 g
Molecular Weight (g/mol) 94.11
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.