Learn More
Sigma Aldrich Fine Chemicals Biosciences 1 way threaded end adapter
Catalog No. 502905192
Encompass
Plated brass or 316 stainless steelMeets ISO 594 specificationsfinal assembly water tested at 45 PSIfemale Luer Lock (fLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.Micro-Mate(R) female Luer Lock (MMfLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.Male Luer (ML) - male Luer slip that fits with a female Luer.
Catalog No. 50-290-5192 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAD633010EA
Plated brass or 316 stainless steelMeets ISO 594 specificationsfinal assembly water tested at 45 PSIfemale Luer Lock (fLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.Micro-Mate(R) female Luer Lock (MMfLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.Male Luer (ML) - male Luer slip that fits with a female Luer.