Learn More
Electron Microscopy Sciences Cardboard Well Slide In Aluminum Holder, 1 Well with 64 Numbered Spaces, 2.5 x 4.5 mm, White
SDP
Catalog No. 501930096
Encompass_Preferred
EMS introduces these unique cardboard "well" slides. Complete with glass covered slide and aluminum holder, these slides are less prone to static as compared to similar plastic slides.
Wells: 1 Well with 64 Numbered Spaces
Size: 2.5 x 4.5 mm
Color: White
Slide: 76 mm x 27.2 mm x 4.5 mm (H)
Catalog No. 50-193-0096 Supplier Electron Microscopy Sciences Supplier No. 6351502
EMS introduces these unique cardboard "well" slides. Complete with glass covered slide and aluminum holder, these slides are less prone to static as compared to similar plastic slides.
Wells: 1 Well with 64 Numbered Spaces
Size: 2.5 x 4.5 mm
Color: White
Slide: 76 mm x 27.2 mm x 4.5 mm (H)