Learn More
CELLTREAT Scientific Products 1 Well Tissue Culture Plate, Sterile
SDP
Catalog No. 501149068
Encompass_Preferred
CELLTREAT Scientific Products 1 Well Tissue Culture Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. 1 Well Plate has the same footprint as standard multiple well plates and is useful for high volume and robotic screening of DNA libraries and manual plating; it is also suitable for dot-blotting bacterial clones on 11.4 x 7.3cm (4.5 x 2.875in) nylon membranes. Has a cell growth area of 97.4cm¿. RNase/DNase Free and non-pyrogenic. Every case is printed wihth the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 50-114-9068 Supplier CELLTREAT Scientific Products Supplier No. 229101
CELLTREAT Scientific Products 1 Well Tissue Culture Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. 1 Well Plate has the same footprint as standard multiple well plates and is useful for high volume and robotic screening of DNA libraries and manual plating; it is also suitable for dot-blotting bacterial clones on 11.4 x 7.3cm (4.5 x 2.875in) nylon membranes. Has a cell growth area of 97.4cm¿. RNase/DNase Free and non-pyrogenic. Every case is printed wihth the lot number for quality traceability. Individually packed sterile, 50/case.