Learn More
Accuflow Systems 1 1/2 Inch Silicone Hose
Supplier:  Accuflow Systems 24P1SWTC24TC24SE96
Encompass_Preferred
1-1/2 Inch ID Platinum Cured Wire Reinforced Silicone Hose 96 Inches OAL
Catalog No. 50-194-4553