Learn More
Accuflow Systems 1 Inch Silicone Hose
Supplier:  Accuflow Systems 16P1SWTC16TC16SE24
Encompass_Preferred
1 Inch ID Platinum Cured Wire Reinforced Silicone Hose 24 Inches OAL
Catalog No. 50-194-4545