Learn More
Gmp Labeling Inc Durable 1 x 0.5 GREEN Therm Trans Lbl
SDP
Catalog No. NC3483017
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 1" x 0.5", Green, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483017 Supplier Gmp Labeling Inc Supplier No. L210TTGRN
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 1" x 0.5", Green, Permanent Ahdesive, 1000 labels/roll