Learn More
Gmp Labeling Inc Durable 1 x 0.5 RED Therm Trans Lbl
SDP
Catalog No. NC3483024
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 1" x 0.5", Red, Permanent Ahdesive, 1000 labels/roll
Catalog No. NC3483024 Supplier Gmp Labeling Inc Supplier No. L210TTRED
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 1" x 0.5", Red, Permanent Ahdesive, 1000 labels/roll