Learn More
Chemglass Life Sciences 1in Diameter X 1/8in Thick Borosilicate Disc
SDP
Catalog No. CG190601
Encompass_Preferred
Plate glass, disc, borosilicate, 1in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.
Catalog No. CG190601 Supplier Chemglass Life Sciences Supplier No. CG190601
Plate glass, disc, borosilicate, 1in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.