Learn More
Hach Company 1 Year Spare Part Kit for 9245-9240 Sodium Analyzer (All Ranges) - Maintenance kit with electrodes, electrolyte, filters, and tubing
Catalog No. NC1974262 Shop All Hach Company Products
Encompass
One-year spare part kit for the Hach 9245-9240 sodium analyzer that supplies replacement electrodes, electrolyte, in-line filters, and tubing to support routine maintenance and reliable sodium measurement. Available for one-time purchase or recurring pick-and-ship orders.
  • Provides essential replacement components for routine analyzer maintenance.
  • Includes reference electrode, sodium glass electrode, electrolyte (500 mL), filters, and replacement tubing.
  • Reduces downtime by supplying commonly needed service parts in a single kit.
  • Eligible for recurring shipment to support scheduled maintenance programs.
  • Component SDS documentation is available for safe handling and compliance.
Catalog No. NC1974262 Supplier Hach Company Supplier No. 09240=A=8000
One-year spare part kit for the Hach 9245-9240 sodium analyzer that supplies replacement electrodes, electrolyte, in-line filters, and tubing to support routine maintenance and reliable sodium measurement. Available for one-time purchase or recurring pick-and-ship orders.
  • Provides essential replacement components for routine analyzer maintenance.
  • Includes reference electrode, sodium glass electrode, electrolyte (500 mL), filters, and replacement tubing.
  • Reduces downtime by supplying commonly needed service parts in a single kit.
  • Eligible for recurring shipment to support scheduled maintenance programs.
  • Component SDS documentation is available for safe handling and compliance.