Learn More
Hampton Research Ammonium formate 200mL, CAS 540-69-2, MFCD00013103, Purity ≥ 99.0%
SDP
Catalog No. 502555
Encompass
10.0 M Ammonium formate - 200 ml. Crystallization grade Ammonium formate for formulating screens or for optimization
Catalog No. 50-255-5 Supplier Hampton Research Supplier No. HR2659
10.0 M Ammonium formate - 200 ml. Crystallization grade Ammonium formate for formulating screens or for optimization