Learn More
Polysciences, Inc. Polybead Polystyrene Microsphere 10.0um, Suited for Protein binding using passive adsorption techniques, 10.0um particles packaged as 2.5% aqueous suspension
SDP
Supplier:  Polysciences, Inc. 171365
Encompass
Polybead Polystyrene Microsphere 10.0um, Suited for Protein binding using passive adsorption techniques, 10.0um particles packaged as 2.5% aqueous suspension, BP: 100 deg.C, MP: 0 deg.C, Store at 4 deg.C
Catalog No. NC9124974