Learn More
Inorganic Ventures 10 000µg/mL CHLORIDE 125mL
SDP
Catalog No. 50272310
Encompass_Preferred
Matrix: Water (H2O), Starting Material: KCl, Analyte: Cl- 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-272-310 Supplier Inorganic Ventures Supplier No. ICCL10125ML
Matrix: Water (H2O), Starting Material: KCl, Analyte: Cl- 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search