Learn More
Anatrace D310-CH210 - 10:1 DDM/CHS Pre-Made Solution, 1ml
Catalog No. NC3769351
Encompass
Dodecyl Maltoside (D310) and Cholesteryl Hemisuccinate (CH210) Solution • DDM/CHS Solution • Dodecyl-β-D-Maltoside (100 mg/ml):CHS-TRIS (10 mg/ml) Solution
Catalog No. NC3769351 Supplier Anatrace Supplier No. D310CH210 1 ML
Dodecyl Maltoside (D310) and Cholesteryl Hemisuccinate (CH210) Solution • DDM/CHS Solution • Dodecyl-β-D-Maltoside (100 mg/ml):CHS-TRIS (10 mg/ml) Solution