Learn More
Cayman Chemical 10 11-Cl-BBQ 10mg
SDP
Catalog No. 502990618
Encompass_Preferred
A mixture of 10-Cl-BBQ and 11-Cl-BBQ that acts as an AhR agonist; induces Cyp1a1 transcription in mouse liver at 10 mg/kg
Catalog No. 50-299-0618 Supplier Cayman Chemical Supplier No. 2997810 MG
A mixture of 10-Cl-BBQ and 11-Cl-BBQ that acts as an AhR agonist; induces Cyp1a1 transcription in mouse liver at 10 mg/kg