Learn More
Cayman Chemical 10 11-dehydro CurvularIn 1mg
SDP
Catalog No. 502986608
Encompass_Preferred
A natural mycotoxin that has cytotoxic activity against select cancer cell lines; activates the heat shock response, blocks TGF-β signaling, and scavenges superoxide anions
Catalog No. 50-298-6608 Supplier Cayman Chemical Supplier No. 218011 MG
A natural mycotoxin that has cytotoxic activity against select cancer cell lines; activates the heat shock response, blocks TGF-β signaling, and scavenges superoxide anions