Learn More
AdipoGen 10,11-Dehydrocurvularin (1 mg)
SDP
Supplier:  AdipoGen AGCN20165M001
Encompass_Preferred
Chemical. CAS: 1095588-70-7. Formula: C16H18O5. MW: 290.3. Isolated from Aspergillus sp. Antibiotic. Antifungal. Nematicidal. Phytotoxic. TGF-beta signaling inhibitor Anticancer compound. Strong activator of the heat shock response. Inhibitor of the appressorium formation of M. grisea
Catalog No. 50-205-3555