Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10,11-Dehydrocurvularin | 21178-57-4 | 1 MG
SDP

Catalog No. 5000420056
Encompass_Preferred

10,11-Dehydrocurvularin is a prevalent fungal phytotoxin and an antibiotic. It acts as a strong activator of the heat shock response, inhibits the TGF-β signalling pathway, and demonstrates anti-tumorous activity.

  • Purity of 99.59%
  • Molecular weight of 290.31
  • Chemical formula: C16H18O5
  • Appears as a solid, white to light yellow
  • Originates from microorganisms, Aspergillus terreus
  • Soluble in DMSO

Catalog No. 50-004-20056 Supplier Medchemexpress LLC Supplier No. HYN6679A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10,11-Dehydrocurvularin is a prevalent fungal phytotoxin and an antibiotic. It acts as a strong activator of the heat shock response, inhibits the TGF-β signalling pathway, and demonstrates anti-tumorous activity.

  • Purity of 99.59%
  • Molecular weight of 290.31
  • Chemical formula: C16H18O5
  • Appears as a solid, white to light yellow
  • Originates from microorganisms, Aspergillus terreus
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.