Learn More
Medchemexpress LLC 10,11-(Methylenedioxy)-20(S)-camptothecin | 135415-73-5 | 99.4% | 392.36 | C21H16N2O6 | 5MG
SDP
Catalog No. 5000239670
Encompass_Preferred
FL118 (10,11-(Methylenedioxy)-20(S)-camptothecin) is a camptothecin analogue research compound that functions as a potent survivin inhibitor with reported oral activity in preclinical studies. It acts as a molecular glue degrader of DDX5 (p68), promoting dephosphorylation and degradation of oncogenic proteins, and is provided as a high-purity material for in vitro and in vivo cancer research.
  • Has potent survivin inhibition and demonstrated antitumor activity.
  • Acts as a molecular glue degrader targeting DDX5 to modulate oncogenic pathways.
  • Suitable for both in vitro and in vivo cancer research applications.
  • Provided at high purity to support experimental reproducibility.
  • Available in small-mass packages appropriate for preclinical studies.
  • Molecular weight 392.36 and formula C21H16N2O6 for accurate dosing calculations.
This item is not returnable. View return policy
Catalog No. 50-002-39670 Supplier Medchemexpress LLC Supplier No. HY124865MG
FL118 (10,11-(Methylenedioxy)-20(S)-camptothecin) is a camptothecin analogue research compound that functions as a potent survivin inhibitor with reported oral activity in preclinical studies. It acts as a molecular glue degrader of DDX5 (p68), promoting dephosphorylation and degradation of oncogenic proteins, and is provided as a high-purity material for in vitro and in vivo cancer research.
  • Has potent survivin inhibition and demonstrated antitumor activity.
  • Acts as a molecular glue degrader targeting DDX5 to modulate oncogenic pathways.
  • Suitable for both in vitro and in vivo cancer research applications.
  • Provided at high purity to support experimental reproducibility.
  • Available in small-mass packages appropriate for preclinical studies.
  • Molecular weight 392.36 and formula C21H16N2O6 for accurate dosing calculations.