Learn More
Cayman Chemical 10,11-dihydro-10-hydroxy Carbamazepine, Purity: ≥98%, CAS: 29331-92-8, 50 mg
SDP
Catalog No. 502968027
Encompass_Preferred
An antiepileptic agent and active metabolite of oxcarbazepine; a racemic mixture of (R)- and (S)-10,11-dihydro-10-hydroxy carbamazepine
Catalog No. 50-296-8027 Supplier Cayman Chemical Supplier No. 1846750 MG
An antiepileptic agent and active metabolite of oxcarbazepine; a racemic mixture of (R)- and (S)-10,11-dihydro-10-hydroxy carbamazepine