Learn More
Cayman Chemical 10 11dIhydro10 11dIhydrxy 10mg
SDP
Catalog No. 502909532
Encompass_Preferred
A metabolite of carbamazepine and oxcarbazepine; has been found in wastewater effluent
Catalog No. 50-290-9532 Supplier Cayman Chemical Supplier No. 3543610 MG
A metabolite of carbamazepine and oxcarbazepine; has been found in wastewater effluent