Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Heptacosadiynoic Acid 98.0+%, TCI America™
SDP

Catalog No. H0666500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0666500MG 500 mg
H0666100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0666500MG Supplier TCI America Supplier No. H0666500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67071-94-7
Molecular Formula C27H46O2
Molecular Weight (g/mol) 402.663
MDL Number MFCD00060065
InChI Key DWPBEQMBEZDLDV-UHFFFAOYSA-N
PubChem CID 20064705
IUPAC Name heptacosa-10,12-diynoic acid
SMILES CCCCCCCCCCCCCCC#CC#CCCCCCCCCC(=O)O

Specifications

Melting Point 69°C
Quantity 500 mg
Formula Weight 402.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Heptacosadiynoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.