Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Heptadecadiynoic Acid 97.0+%, TCI America™
SDP

Catalog No. H0775100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0775100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0775100MG Supplier TCI America Supplier No. H0775100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28393-06-8
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.39
MDL Number MFCD00142348
InChI Key CLTRJQSBWDSZHN-UHFFFAOYSA-N
PubChem CID 44630341
IUPAC Name heptadeca-10,12-diynoic acid
SMILES CCCCC#CC#CCCCCCCCCC(O)=O

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 100 mg
Formula Weight 262.39
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Heptadecadiynoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.