Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Pentacosadiyn-1-ol 98.0+%, TCI America™
SDP

Catalog No. P10531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10531G 1 g
P1053100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10531G Supplier TCI America Supplier No. P10531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92266-90-5
Molecular Formula C25H44O
Molecular Weight (g/mol) 360.626
MDL Number MFCD00059096
InChI Key XHKKFKTVKSZQKA-UHFFFAOYSA-N
PubChem CID 5122328
IUPAC Name pentacosa-10,12-diyn-1-ol
SMILES CCCCCCCCCCCCC#CC#CCCCCCCCCCO

Specifications

Melting Point 60°C
Color White
Quantity 1 g
Formula Weight 360.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Pentacosadiyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.