Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 10,12-Pentacosadiynoic acid

Catalog No. 764925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-492-5G 5 g
76-492-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-492-5G Supplier Sigma Aldrich Supplier No. 764925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 10,12-pentacosadiynoic acid (pcda) is an amphiphilic diacetylene that can be used:
  • In combination with a semifluorinated tetracosane ((cf3(cf2)7(ch2)15ch3) to synthesize mixed monolayer by photopolymerization
  • For the conversion of hydrophobic upconverting nanoparticles (ucnps) to hydrophilic
  • To synthesize a polymer by combining with pyridine-2-aldoxime for colorimetric detection of malathion
  • To synthesize a thermochromic sensor constituting polymer fibres with monomeric unit of two pcda linked by a p-phenylene group (bis-pcda-pH)
  • As an inert coating material upon self-assembly and photopolymerization to modify the surface of fe3o4 nanoparticles

Specifications

CAS 66990-32-7
Molecular Formula C25H42O2
Linear Formula C25H42O2
MDL Number MFCD00041684
Quantity 5 g
Synonym PCDA
Molecular Weight (g/mol) 374.6
Percent Purity ≥97.0% (HPLC)
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.