Learn More
Ace Glass, Inc. 10-12mm tubing clamps, Box of 100
SDP
Catalog No. 509558909
Encompass
10-12mm tubing clamps eliminate older method of wiring rubber tubing to glass. Requires only one hand to apply, and can be instantly applied or removed. Box of 100.
Catalog No. 50-955-8909 Supplier Ace Glass, Inc. Supplier No. 1114520
10-12mm tubing clamps eliminate older method of wiring rubber tubing to glass. Requires only one hand to apply, and can be instantly applied or removed. Box of 100.