Learn More
Medchemexpress LLC HY-128976 1mg Medchemexpress, 10β,17β-dihydroxyestra-1,4-dien-3-one CAS:549-02-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1289761MG
Encompass_Preferred
Medchemexpress, HY-128976 1mg 10β,17β-dihydroxyestra-1,4-dien-3-one CAS:549-02-0 10β,17β-dihydroxyestra-1,4-dien-3-one (DHED) is a brain-targeting bioprecursor prodrug of the main human estrogen, 17β-estradiol, alleviates hot flushes in rat models of thermoregulatory dysfunction of the brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9789