Learn More
Ace Glass, Inc. Thermometer adapter, PTFE, 10/18 to compression seal, use with 6-6.5mm diameter tubes, Viton; O-ring
SDP
Catalog No. NC9040591
Encompass_Preferred
10/18 PTFE thermometer adapter with FETFE o-ring, for use with plain stem thermometers, gas inlet tubes, etc. The o-ring compression seal on the adapter allows adjustable depth positioning. Adapter accomm ODates tubes up to 6.5mm.
Catalog No. NC9040591 Supplier Ace Glass, Inc. Supplier No. 830005
10/18 PTFE thermometer adapter with FETFE o-ring, for use with plain stem thermometers, gas inlet tubes, etc. The o-ring compression seal on the adapter allows adjustable depth positioning. Adapter accomm ODates tubes up to 6.5mm.