Learn More
Chemglass Life Sciences 10/18 Outer Joint, Dewar Seal, 30mm OD Flare
SDP
Catalog No. CG13130
Encompass_Preferred
10/18 Outer Joint, Dewar Seal, 30mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.
Catalog No. CG13130 Supplier Chemglass Life Sciences Supplier No. CG13130
10/18 Outer Joint, Dewar Seal, 30mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.