Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-10,2-Camphorsultam 98.0+%, TCI America™
SDP

Catalog No. C13251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13251G 1 g
C13255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13251G Supplier TCI America Supplier No. C13251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94594-90-8
Molecular Formula C10H17NO2S
Molecular Weight (g/mol) 215.311
MDL Number MFCD00066271
InChI Key DPJYJNYYDJOJNO-SZBHIRRCSA-N
Synonym 5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, --10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
PubChem CID 54579826
SMILES CC1(C2CCC13CS(=O)(=O)NC3C2)C

Specifications

Melting Point 182°C
Color White
Quantity 1 g
Formula Weight 215.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-10,2-Camphorsultam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.