Learn More
Thermco Products Inc Precision Taper Joint Non-Hg thermometer, Spirit, -10 deg./250 deg.C, 1 deg.C div, 25mm imm, 315mm, 10/18 taper
Catalog No. NC0591819
Encompass_Preferred
Manufactured with a 10/30 glass standard taper joint to fit in 10/18 joint of flasks and other laboratory glassware. Accurate to N.I.S.T. tolerances. Individually serialized. Thermometer may be Safety-Coated with TFE. Filled with a safe red spirit filling for safety purposes.
Catalog No. NC0591819 Supplier Thermco Products Inc Supplier No. ACCST1801250S
Manufactured with a 10/30 glass standard taper joint to fit in 10/18 joint of flasks and other laboratory glassware. Accurate to N.I.S.T. tolerances. Individually serialized. Thermometer may be Safety-Coated with TFE. Filled with a safe red spirit filling for safety purposes.