Learn More
MSC Baldwin Filters Air Filter, 4-3/32 x 10-29/32 in, 1 ea
Catalog No. 5000505720
Encompass
This inner air element is designed for automotive engines. It functions as an engine inner air filter, preventing airborne contaminants from reaching the combustion chamber.
  • Engine inner air filter type
  • Round design
  • Axial seal filters have securely affixed metal end caps with a raised gasket for proper sealing
  • Media material: Cellulose, paper
  • Overall height: 10 29/32 in
  • Outside diameter: 4 3/32 in
  • Inside diameter: 21/32 in
  • Outside diameter - bottom: 3 17/32 in
  • Inside diameter - bottom: 3 17/32 in
Catalog No. 50-005-05720 Supplier MSC Supplier No. 10938785
This inner air element is designed for automotive engines. It functions as an engine inner air filter, preventing airborne contaminants from reaching the combustion chamber.
  • Engine inner air filter type
  • Round design
  • Axial seal filters have securely affixed metal end caps with a raised gasket for proper sealing
  • Media material: Cellulose, paper
  • Overall height: 10 29/32 in
  • Outside diameter: 4 3/32 in
  • Inside diameter: 21/32 in
  • Outside diameter - bottom: 3 17/32 in
  • Inside diameter - bottom: 3 17/32 in